Order Entry
Finland
ContactUsLinkComponent
 
Biebrich scarlet
Catalog # CHRO1A-398
Supplier:  CHROMA
CAS Number:  
Biebrich scarlet
Catalog # CHRO1A-398
Supplier:  CHROMA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :556.49 g/mol
Storage Temperature:Ambient
EINECS:224-084-5
CAS Number:4196-99-0
Formula:C₂₂H₁₄N₄Na₂O₇S₂
MDL Number:MFCD00003891