Order Entry
Finland
ContactUsLinkComponent
Iridium standard solution, 1,000 mg/L Ir in 10% hydrochloric acid (from IrCl₃) standard for AAS and ICP
Iridium standard solution, 1,000 mg/L Ir in 10% hydrochloric acid (from IrCl₃) standard for AAS and ICP
Catalog # PERCN9300125
Supplier:  PerkinElmer
CAS Number:  
Iridium standard solution, 1,000 mg/L Ir in 10% hydrochloric acid (from IrCl₃) standard for AAS and ICP
Catalog # PERCN9300125
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HCl
  • Pk:
    1
  • Element:
    Iridium
  • Symbol:
    Ir

Specifications

About this item