Transfer tubes, Transfertube®
Supplier: Spectrum Laboratories
Certificates
About this item
Disposable tubes for liquid and solid samples with one hand. The sample to be transfered is inserted with harvester and discharged using the plunger.
- Quick and easy use with the finger ring
- Reduces the risk of cross contamination
Recommended for particulates, suspensions, and microbial colonies.