Corning® Interleukin-3 (IL-3) Culture Supplement, Corning
Catalog # 734-2390
Supplier: Corning
CAS Number:
Specifications
- Pk:25 mL
- Description:Interleukin- 3 (IL-3) culture supplement, mouse
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO 14001 - Bedford
Specifications
About this item
Interleukin-3 (IL-3) is a glycoprotein that regulates differentiation and enhances proliferation of a variety of hematopoietic precursor cells, mast cells, basophils, and NK cells.
IL-3 Culture Supplement, mouse, is used to culture a variety of IL-3 responsive cells, e.g., mast cells, basophils, natural killer cells, and several hematopoietic precursor cells.
Mouse supplement is tested for ability to promote proliferation of an IL-3 dependent mouse cell line (32-DCL) in serum-containing medium. Mouse recombinant supplement is shown to stimulate proliferation of IL-3 dependent murine cells (FDC-P1). All products are filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.