Powder samplers, MicroMate
The MicroMate is designed for small volumes of powders and granules.
- Inner rod extends beyond the outer tube
- Slightly variable nominal sample volume
- Dry and ambient recommended storage temperature
Mostly used in unit dosing in the pharmaceutical industries.