Clinical swabs (Sterilin)
: Sterilin
This pre-labelled tube with liquid medium and flocked swab can be processed using laboratory procedures for bacterial culture and molecular detection of bacteria, viruses and Chlamydia.
- Medium enabling multi-test analysis
- Free of enzymes and inhibitors
- May interfere with nucleic acid amplification assays
Designed for the collection and transport of clinical specimens containing aerobes, anaerobes, fastidious bacteria, viruses and Chlamydia.
: E-Swab MRSA kits contain multiple swabs for ease of sample collection and processing.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904