Buffers, concentrated solutions, Titrisol®, Supelco®
Supplier: Merck
Certificates
About this item
Titrisol® buffer concentrate for 500 ml buffer solution [20 °C].
- Traceable to secondary reference material (SRM) of NIST and PTB
- Buffer concentrate for the preparation of 500 ml of buffer solution
- Accuracy: ≤0,05 pH units at 20 °C
* according to Wiese for 1000 ml solution
Limited Time Sale
Select items below are currently on sale
Specifications
- M-Clarity quality level:MQ100