Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trays/Dishes/Pots
Laboratory Trays/Dishes
Dishes and lids
Dishes and lids
Dishes and lids
Supplier:
NICKEL ELECTRO
Certificates
Expand image
Dishes and lids
Supplier:
NICKEL ELECTRO
About this item
Made of aluminium.
Very good temperature resistance
Good chemical resistance
Lightweight