Crucibles, stainless steel
These stainless steel 304 crucibles with bright finish are much harder and last longer than porcelain types.
- Non-magnetic
- Shatterproof
: Lids need to be ordered separately.
These stainless steel 304 crucibles with bright finish are much harder and last longer than porcelain types.