Taper fittings for standard glass joints
Supplier: COWIE
Certificates
About this item
Compression fittings are manufactured from pure PTFE with a PEEK olive to give a secure grip.
- Chemically inert
- Reduce contamination and simplify cleaning
- Non-scratch
Compression fittings fit standard taper ground glass joints to hold items such as temperature probes and sampling tubes securely in place.