Tubing, PTFE
Supplier: COWIE
Certificates
About this item
PTFE, white.
- Excellent chemical resistance
- Very good thermal resistance, autoclavable
- Free of extractable substances, physiologically neutral, biocompatible
This tubing is characterised by its non stick surface and excellent dielectric properties.
Temperature resistance: −200 to +260 °C