Tiamiinimonofosfaattiklorididihydraatti, EMPROVE® api suitable for use as an active pharmaceutical ingredient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:273724-21-3
:
Formula:C₁₂H₁₈ClN₄O₄PS·2H₂O
MDL Number:MFCD00063473
EINECS:208-536-9
Frequently Bought Together