Polyethylene glycol dimethylether, M.W. 2,000 for synthesis, Sigma-Aldrich®
Catalog # 8.14172.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:49 - 53 °C
- Molar Mass:2000 g/mol
- Boiling Point:250 °C (1013 hPa)
- Vapor Pressure:- 0.01 hPa (20 °C)
- Flash Point:254 °C
- Refractive Index:1.455 (60 °C, 589 nm)
- Density:1.08 g/cm3 (60 °C)
- pH:(100 g/l, H2O, 20 °C) neutral
- Ignition Point:385 °C DIN 51794
- Viscosity kinematic:30 mm2/s (100 °C)
Specification Test Results
About this item
:
:M-Clarity™ quality level = MQ200
CAS Number:24991-55-7
Formula:H₃C(OCH₂CH₂)nOCH₃
MDL Number:MFCD00147972