0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Agarose
Agarose
Agarose
Agarose
Supplier:
PanReac AppliChem
Certificates
Expand image
Agarose
Supplier:
PanReac AppliChem
About this item
CAS Number:
9012-36-6
:
MDL Number:
MFCD00081294
Melting Pt:
60…90 °C
Storage Temperature:
Ambient
EINECS:
232-731-8