Order Entry
Finland
ContactUsLinkComponent
Earmuffs for welding helmets, Peltor™ H505B
Earmuffs for welding helmets, Peltor™ H505B
Catalog # MMMAH505B
Supplier:  3M
undefined
Earmuffs for welding helmets, Peltor™ H505B
Catalog # MMMAH505B
Supplier:  3M
Specifications
  • SNR:
    26 dB
  • Colour:
    Black
  • Description:
    Peltor™ H505B welding helmet earmuffs with neck strap, suitable for use with a welding helmet
  • Pk:
    1
Specifications
About this item
These welding helmet earmuffs with neck strap have been designed for use with welding shields, particularly the 3M™ Speedglass™ 9100.
  • Soft earcups filled with fluid for enhanced comfort
  • The design of the neckband improves compatibility with other personal protective equipment
  • Generous space underneath the earmuffs helps to reduce build-up of heat and sweat
  • Soft, wide earcups guarantee optimum seal and low pressure for outstanding comfort
  • Cushions and inserts are easy to replace and to keep clean and hygienic
The earmuffs provide moderate insulation, which is sufficient for most welding applications.
SNR: 26 dB
Certifications: EN 352-1:2002