Rain overalls with bib and braces, Bandung 6620
: SIOEN
Bib and brace trousers with elastic braces with PVC buttons and buckles. Made of lightweight Flexothane® Essential, 100% polyester knitting with PU coating.
- Waterproof and windproof
- Supple, elasticated material: Very comfortable to wear
- Lightweight and noiseless
- Legs adjustable by press studs
Weight: 170 g/m²
: ENV 343 (3-1)
- Catalog No:
- 113-07147
- 113-07138
- 113-07140
- 113-07146
- 113-07145
- 113-07155
- 113-07144
- 113-07154
- 113-07150
- 113-07151
- 113-07153
- 113-07141
- 113-07152
- 113-07149
- 113-07142
- 113-07148
- 113-07143
- 113-07139
- Model:
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Bandung 6620 Flexothane Essential
- Garment application:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Fabric:
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Lightweight Flexothane® Essential, 100% polyester knitting with PU coating
- Weight:
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²