Chemical resistant rain overalls, Montreal
: SIOEN
Water- and windproof overall made of Flexothane® Classic: 100% polyamide knitting with PU coating. The durable garment offers protection against liquid chemicals.
- Straight collar with foldaway hood
- Zip closure under double flap with press studs
- Raglan sleeves with elasticated wind cuffs, sleeve and ankle narrowing by press studs
- Elasticated waist in the back
- High frequency welded seams
Weight: 180 g/m²
: EN 14786, EN 14605 (Type 4), EN 14126 (Type 4-B), EN 343 (3-1)
Oeko-Tex® Standard 100
Oeko-Tex® Standard 100
- Catalog No:
- 113-07133
- 113-07134
- 113-7688
- 113-7677
- 113-7664
- 113-7678
- 113-7679
- 113-7665
- 113-7666
- 113-7680
- 113-07132
- 113-7667
- 113-7668
- 113-7669
- 113-07129
- 113-07128
- 113-7689
- 113-7681
- 113-7670
- 113-7671
- 113-7682
- 113-7672
- 113-7683
- 113-7673
- 113-7674
- 113-7684
- 113-7685
- 113-7675
- 113-7686
- 113-7676
- 113-7687
- 113-07131
- 113-07135
- 113-07130
- 113-5951
- 113-07136
- Model:
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Montreal
- Garment application:
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Weight:
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²