Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bradford's reagent
Bradford's reagent for quantitative protein determination
Bradford's reagent for quantitative protein determination
Bradford's reagent for quantitative protein determination
BIOZEXP-BFU05L
:
Biozol
:
Bradford's reagent for quantitative protein determination
BIOZEXP-BFU05L
:
Biozol
:
Pack type:
Glass bottle
Pk:
500 mL
Seq:
2725
:
:
GHS08
:
Refrigerator