Buffer, PBS solution 10X concentrate (phosphate buffered saline)
PBS buffer solution 10X concentrate (phosphate buffered saline), Glass bottle, 500 mL
Catalog # EURLECM4053XL
Supplier: EUROCLONE
Buffer, PBS solution 10X concentrate (phosphate buffered saline)
Catalog # EURLECM4053XL
Supplier: EUROCLONE
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Description:PBS buffer solution 10X concentrate (phosphate buffered saline)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



