Dextran sulphate sodium salt, M.W. ~ 500,000 ≥98%, Ultra Pure Grade
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:9011-18-1
:
Formula:(C₆H₇Na₃O₁₄S₃)n
MDL Number:MFCD00081551
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- DNase:none detected
- Heavy Metals:= 0.005 %
- RNase:none detected
- Iron:= 0.001 %
- Loss on Drying:= 10 %
- pH (1%, Water) @25°C:6.4 - 8
- Protease:none
- Purity:98 %