Dextran sulphate sodium salt
Catalog # MOLE38417956-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
CAS Number:9011-18-1
Formula:(C₆H₇Na₃O₁₄S₃)n
MDL Number:MFCD00081551