G418
1 KIT
Catalog # EURLEMR143005
Supplier: EUROCLONE
Jump to:
Product Details & Documents
Synonyms: G-418, (2R,3S,4R,5R,6S)-5-Amino-6-(((1R,2S,3S,4R,6S)-4,6-diamino-3-(((2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-2-((R)-1-hydroxyethyl)tetrahydro-2H-pyran-3,4-diol
- CAS Number:49863-47-0
- Formula:C₂₀H₄₀N₄O₁₀
- MDL Number:MFCD30541267
- Molecular Weight:496.552 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pk:1 KIT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



