Order Entry
Finland
ContactUsLinkComponent
VWR® Terrific, Broth, powder
VWR® Terrific, Broth, powder
Catalog # J869-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Terrific, Broth, powder
Catalog # J869-500G
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    500 g
  • Standards:
    -
  • Description:
    Terrific broth, powder
  • Seq:
    02
Specifications
About this item
Terrific broth is an enriched medium, ideal for culturing cells for nucleic acid purification and recombinant protein expression.
  • Save time using pre-mixed media
  • Homogenous and high quality powders
Terrific broth is an enriched medium containing high levels of tryptone and yeast extract, which provide amino acids, vitamins and trace elements necessary for high density growth. Glycerol is added as a carbohydrate source, while the potassium phosphate is present to both maintain optimal pH and serve as a source of potassium for cellular metabolism. Glycerol is not supplied with this media.
1 L of prepared media contains 12 g of tryptone, 24 g of yeast extract, 9,4 g of potassium phosphate dibasic, and 2,2 g of potassium phosphate monobasic.