Tubing, Viton (FPM)
Supplier: DEUTSCH NEUMANN
Certificates
About this item
Fluororubber (FPM), black.
- Temperatur resistance: −10 to +200 °C
- Hardness: 75 Shore A
- Dimension tolerances according to DIN ISO 3302-1 E2
Good resistance against ozone and climatic exposure as well as gasoline and mineral oil.