Filter adapters for syringes, PTFE, BOLA
Supplier: Bohlender
Certificates
About this item
Adapters can be screwed together into multi-stage filters (such as prefilter, main filter). The low weight easily allows exchange of the optionally available filtering membranes.
- Temperature resistance: –200 to +250 °C
Certifications: FDA conform.