N-Benzylglycine ethyl ester 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6436-90-4
:
Formula:C₁₁H₁₅NO₂
MDL Number:MFCD00009174
:193.25 g/mol
Boiling Pt:140…142 °C (10 mmHg)
Density:1.03 g/cm³
Flash Pt:112 °C
Storage Temperature:Refrigerator
EINECS:229-218-6
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5045 to 1.5065 (20°C, 589 nm)
- GC:≥95 %