Order Entry
Finland
Orders LinkContactUsLinkComponent
Amitraz

Amitraz

Glass bottle for solids, 200 mg
Catalog # PROOBP015
Amitraz
Catalog # PROOBP015
CAS Number:  33089-61-1
Amitraz

Glass bottle for solids, 200 mg

CAS Number:  33089-61-1
Price
500,00 €
Log in for availability
1 * 200 mg
Product Details & Documents
  • CAS Number:33089-61-1
  • Formula:C₁₉H₂₃N₃
  • MDL Number:MFCD00069396
  • EINECS:251-375-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    200 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.