



Hydrogen chloride 4 N in 1,4-dioxane, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
:36.46 g/mol
Flash Pt:17 °C
Storage Temperature:Ambient
EINECS:231-595-7
UN:2924
ADR:3,II
:GHS07
:GHS08
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Solution
- Strength:3.95 to 4.40 N