Order Entry
Finland
Orders LinkContactUsLinkComponent
2-(4-Nitrophenyl)ethanol

2-(4-Nitrophenyl)ethanol

Glass bottle for solids, 5 g
Catalog # MOLEM77045227
Marketsource
Supplier:   Molekula
2-(4-Nitrophenyl)ethanol
Catalog # MOLEM77045227
Supplier:  Molekula
Marketsource
CAS Number:  100-27-6
2-(4-Nitrophenyl)ethanol

Glass bottle for solids, 5 g

CAS Number:  100-27-6
Price
77,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  4-Nitrophenethyl alcohol, p-Nitrophenethyl Alcohol
  • CAS Number:100-27-6
  • Formula:C₈H₉NO₃
  • MDL Number:MFCD00010202
  • Molecular Weight:167.16 g/mol
  • EINECS:202-835-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.