4-Bromo-2-thenoic acid
Catalog # MAYBCC28501DA
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:207.05 g/mol
CAS Number:16694-18-1
Formula:C₅H₃BrO₂S
MDL Number:MFCD03422294