Pipette aid, SP Bel-Art
Catalog # BELAF378860000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Colour:Blue/white
- Description:Pipette aid for pipettes, Vikem® viny bulb with PE chuck, Hר: 100×50 mm
- Seq:02
- Pk:6
Specifications
About this item
This pipette aid is used by squeezing, then releasing the bulb to aspirate fluid into the pipette.
- Quickly cover pipette top with a gloved finger to control and dispense fluid
- Vikem® vinyl bulb has a tapered polyethylene chuck for quick removal when pipette is filled