Order Entry
Finland
ContactUsLinkComponent
Mini tongs, Sterileware®, SP Bel-Art
Mini tongs, Sterileware®, SP Bel-Art
Catalog # BELAF379440000
undefined
Mini tongs, Sterileware®, SP Bel-Art
Catalog # BELAF379440000
Specifications
  • Length:
    110 mm
  • Pk:
    25
  • Type:
    Sterile
Specifications
About this item
PS, white. For handling food, biological, chemical or pharmaceutical samples.
  • Individually packaged, lot traceable and sterilized sample handling tongs
  • Molded from smooth white polystyrene
  • Serrated jaws securely hold small quantities of moist or dry material
  • Disposable, designed for one time use to avoid cross contamination
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904