Order Entry
Finland
ContactUsLinkComponent
Laboratory glass cleaner, borer 21 SOLID
Laboratory glass cleaner, borer 21 SOLID
Catalog # 148-5191
Supplier:  Borer Chemie
undefined
Laboratory glass cleaner, borer 21 SOLID
Catalog # 148-5191
Supplier:  Borer Chemie
Specifications
  • Description:
    borer 21 SOLID
  • Pk:
    10 kg
Specifications
About this item
Mildly alkaline cleaning powder for automated cleaning of laboratory glassware und reusable laboratory material.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • With a broad cleaning spectrum and excellent results even at low concentrations
  • High cleaning performance due to excellent dispersing capabilities
  • Good material compatibility, especially recommended for alkali and corrosion sensitive items, also suitable for aluminium
  • EDTA-, NTA-, chlorine- and surfactant-free, no release of surfactants into waste water
  • Rinsing with an acidic deconex® cleaner is recommended as the completion of the cleaning process