
2-Methyl-1-tetralone 98%
Catalog # ACRO172370025
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 g
- Seq:3122
Specifications
Specification Test Results
- Appearance:Clear yellow to orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5525 to 1.5545 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
About this item
:
:160.22 g/mol
Boiling Pt:127…131 °C (12 mmHg)
Density:1.06 g/cm³
Flash Pt:>110 °C
EINECS:216-461-8
:GHS07
Storage Temperature:Ambient
CAS Number:1590-08-5
Formula:C₁₁H₁₂O
MDL Number:MFCD00001690