Order Entry
Finland
ContactUsLinkComponent
(2-Naphthoxy)acetic acid for synthesis, Sigma-Aldrich®
Irritant
(2-Naphthoxy)acetic acid for synthesis, Sigma-Aldrich®
Catalog # 8.41023.0100
Supplier:  Merck
CAS Number:  
Irritant
(2-Naphthoxy)acetic acid for synthesis, Sigma-Aldrich®
Catalog # 8.41023.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Melting Point:
    155 - 157 °C
  • Molar Mass:
    202.21 g/mol
  • Hill Formula:
    C12H10O3

Specification Test Results

About this item

 :
 :202.21 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:204-380-0
 :GHS07
Storage Temperature:Ambient
CAS Number:120-23-0
Formula:C₁₂H₁₀O₃
MDL Number:MFCD00004066