Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-5-formylpyridine
2-Chloro-5-formylpyridine 96%
2-Chloro-5-formylpyridine 96%
:
Thermo Fisher Scientific
2-Chloro-5-formylpyridine 96%
:
Thermo Fisher Scientific
:
23100-12-1
:
:
C₆H₄ClNO
:
MFCD03095223
:
141.56 g/mol
:
77…81 °C
:
GHS07
:
Ambient
msds_ALFAH55550
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years