
Diethylene glycol monoethyl ether ≥99.0% (by GC)
: TCI
:111-90-0
:
:C₆H₁₄O₃
:MFCD00002872
:134.18 g/mol
:14,01800
:196°C
:–80 °C
:0,99 g/cm³ (20 °C)
:96°C
:Ambient
:203-919-7
:01-2119475105-42
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %