Order Entry
Finland
ContactUsLinkComponent
 
Mercury standard solution, 1,000 mg/L Hg in 10% nitric acid standard for ICP-MS
  ACCUICPMS34N01X1
 :  
Mercury standard solution, 1,000 mg/L Hg in 10% nitric acid standard for ICP-MS
  ACCUICPMS34N01X1
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Mercury
  • Symbol:
    Hg