2-Cyclohexen-1-one ≥96.0% (by GC)
: TCI
:930-68-7
:
:C₆H₈O
:MFCD00001577
:96.13 g/mol
:171°C
:0,99 g/cm³ (20 °C)
:56°C
:213-223-5
:2929
:6.1,II
:GHS02
:GHS06
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 96.0 %