Cesium standard solution, 1,000 mg/L Cs in dil. nitric acid standard for ICP-MS
Catalog # ACCUICPMS12N0.1X1
Supplier: AccuStandard
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2-5% HNO₃
- Pack type:Glass bottle
- Pk:100 mL
- Element:Cesium
- Symbol:Cs