Order Entry
Finland
ContactUsLinkComponent
Plates for isolators and cleanrooms, Envirocheck® Settle
Plates for isolators and cleanrooms, Envirocheck® Settle
Catalog # 1.18410.0020
Supplier:  Merck
CAS Number:  
undefined
Plates for isolators and cleanrooms, Envirocheck® Settle
Catalog # 1.18410.0020
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    20
  • Standards:
    -
  • Description:
    Tryptic soy agar with neutralisers (0,07% lecithin; 0,5% polysorbate 80; 0,05% histidine; 0,05% thiosulphate)
  • Filling:
    25 ml

Specifications

About this item

There is increasing need for effective air monitoring in all areas where airborne microorganisms may contaminate or affect industrial products and processes. Air monitoring is, therefore, highly important in all industries which have cleanroom areas with filtered air. The Envirocheck® Settle plates (90 mm diameter) can easily be used for active air sampling in combination with the different MAS-100® microbial air monitoring systems. In addition, the plates can also be used for passive air sampling. To fulfil the requirements for protected areas the Envirocheck® Settle plates are triple-wrapped and gamma-irradiated. The 90 mm Settle plates are filled with 25 ml of media to avoid desiccation, for example, after active air sampling of 1000 litres and long incubation of up to 7 days. The inner bag is impermeable to H₂O₂ and includes a pouch containing a drying agent to minimise the formation of condensation. All plates are stored at room temperature (15 to 25 °C), hence refrigeration is not required. Storage at site of application is possible.

  • Gamma-irradiated, triple-wrapped, H₂O₂-impermeable inner bag
  • Fill volume of 25 ml enable long incubation times
  • Four neutralisers to inactivate any residual disinfectants
  • A pouch containing a drying agent minimises the formation of condensation
  • Product name, expiry date, lot number and serial production number are imprinted on each Envirocheck® Settle plate for safe and easy product identification and shelf life monitoring
Packaging: Triple-wrapped, 2 sets of 10 plates each.