Order Entry
Finland
ContactUsLinkComponent
Isopropanol (Propan-2-ol), sterile spray
Isopropanol (Propan-2-ol), sterile spray
Catalog # 148-0025
Supplier:  Texwipe
undefined
Isopropanol (Propan-2-ol), sterile spray
Catalog # 148-0025
Supplier:  Texwipe

Specifications

  • Description:
    2-Propanol 70% (v/v) spray
  • Seq:
    01
  • Packed:
    473 ml (16 fl.oz.) trigger spray bottle
  • Pk:
    12

Specifications

About this item

Sterile 70% Propan-2-ol solution, pre-filtered.

  • Fully assembled trigger spray delivers fine stream or coarse spray
  • Gamma irradiated to a SAL (Sterility Assurance Level) of 10⁻⁶
  • With lot specific certificates of irradiation and analysis
Packaging: Double-bagged and packaged in sealed polybag case liner.