Filtration crucibles, jacketed, VitraPOR®
Supplier: ROBU GLASFILTER
Certificates
About this item
The crucible is made of pure borosilicate glass 3.3, versatile and recyclable material offers excellent properties for the most diverse fields.
- Porosity classes ranges from 1,0 to 500 µm
- Contains heating mantle and standard joints
- Highly resistant to most chemicals such as solvents, acids and alkalis