Filter candles, cylindrical, VitraPOR®
Supplier: ROBU GLASFILTER
Certificates
About this item
These cylindrical filter candles are available with or without tube.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion
Made of borosilicate glass 3.3.