Micro filter candles, VitraPOR®
Supplier: ROBU GLASFILTER
Certificates
About this item
These cylindrical micro filter candles are available with or without tube.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion
Made of Borosilicate glass 3.3.