Order Entry
Finland
ContactUsLinkComponent
Coliwasa trumpet, Wheaton®
Coliwasa trumpet, Wheaton®
Catalog # WHEA885235
Supplier:  DWK Life Sciences
undefined
Coliwasa trumpet, Wheaton®
Catalog # WHEA885235
Supplier:  DWK Life Sciences
Specifications
  • Capacity:
    200 ml
  • Description:
    Disposable samplers
  • Length:
    102 cm
  • Material:
    Glass
  • Pk:
    12
Specifications
About this item
This sampler is designed for the extraction of sludge-type or semi-solid materials.
Suitable for boring through materials.