Büchner funnels, single piece, SP Bel-Art
HDPE, with Fritware® porous plates.
- Can be used with or without filter paper
- For use below 52 °C
- To be used with Vac-Ring™ filter seal for quick filter set up
These fast flowing funnels with sealed-in PE filter plate of coarse porosity (90 to 130 µm) offer good retention and fast flow under vacuum.