Stirring shafts, two paddles, BOLA
Stirrer shafts made of PTFE-jacketed stainless steel and the paddles made of PTFE.
- Chemical resistant
- Temperature resistance from –200 to +250 °C
Two paddles arranged crosswise at 90°. Upper paddle is fixed by means of clamp screws made of PEEK compound.