Ground joint tube fittings, BOLA
Supplier: Bohlender
Certificates
About this item
PTFE, white.
- Temperature resistance: −200 to +250 °C
- Universal chemical resistance
- Autoclavable
For connecting tubes, hard-walled tubing, thermometers. The sealing rings on the outside of the cone prevent sticking of the ground joints and improve the sealing. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.