Folic acid test
Folic acid test, 1 KIT
Catalog # 709-0265
Supplier: TECRA
Jump to:
Product Details & Documents
Simple test in microtitre format for measuring folic acid in raw and fortified foodstuffs.
- Fast: results in less than two hours
- A large number of samples can be tested simultaneously
- For 14 up to a maximum of 43 tests per plate
- Simple interpretation of the results
Specifications
Specifications
- Pk:1 KIT
- Description:Folic acid test